Assay Details
| Gene Symbol: CCL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032852.1 | NP_001028024.1 | 65 | 78 | ||
| XM_015126152.2 | XP_014981638.1 | 613 | 78 | |||
| GenBank mRNA | AF449271.1 | - | 65 | 78 | ||
| JU476728.1 | - | 124 | 78 | |||
More Information
Set Membership: |
|

CCL17
Genomic Map