Assay Details
| Gene Symbol: COX7A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040278.2 | NP_001035368.1 | 276 | 76 | ||
| XM_015123564.2 | XP_014979050.1 | 903 | 76 | |||
| GenBank mRNA | AY585862.1 | - | 229 | 76 | ||
| JU323488.1 | - | 270 | 76 | |||
| JV045060.1 | - | 276 | 76 | |||
| JV707156.1 | - | 89 | 76 | |||
More Information
Set Membership: |
|

COX7A1
Genomic Map