Assay Details
| Gene Symbol: LPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028847001.1 | XP_028702834.1 | 4399 | 58 | ||
| GenBank mRNA | J04635.1 | - | 2485 | 58 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LPA| Gene Symbol: LPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028847001.1 | XP_028702834.1 | 4399 | 58 | ||
| GenBank mRNA | J04635.1 | - | 2485 | 58 | ||
Set Membership: |
|