Assay Details
| Gene Symbol: LPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028847001.1 | XP_028702834.1 | 3054 | 62 | ||
| GenBank mRNA | J04635.1 | - | 1140 | 62 | ||
| JV544878.1 | - | 98 | 62 | |||
More Information
Set Membership: |
|

LPA
Genomic Map