Assay Details
| Gene Symbol: ZCCHC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195739.1 | NP_001182668.1 | 658 | 151 | ||
| XM_015140220.2 | XP_014995706.2 | 736 | 151 | |||
| XM_015140222.2 | XP_014995708.2 | 685 | 151 | |||
| GenBank mRNA | CO647619.1 | - | 664 | 151 | ||
| DQ148059.1 | - | 71 | 151 | |||
| JU336685.1 | - | 596 | 151 | |||
More Information
Set Membership: |
|

ZCCHC9
Genomic Map