Assay Details
Gene Symbol: RAB11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261484.1 | NP_001248413.1 | 106 | 84 | ||
GenBank mRNA | JU337099.1 | - | 140 | 84 | ||
JU477835.1 | - | 106 | 84 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
