Assay Details
Gene Symbol: ARGLU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194022.1 | NP_001180951.1 | 813 | 78 | ||
GenBank mRNA | JU335222.1 | - | 672 | 78 | ||
JU335223.1 | - | 672 | 78 | |||
JU476185.1 | - | 683 | 78 | |||
JU476186.1 | - | 683 | 78 | |||
JU476187.1 | - | 683 | 78 | |||
JV047495.1 | - | 749 | 78 | |||
JV585921.1 | - | 655 | 78 | |||
JV636510.1 | - | 993 | 78 | |||
JV688400.1 | - | 802 | 78 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
