Assay Details
| Gene Symbol: RNF126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261265.1 | NP_001248194.1 | 756 | 101 | ||
| XM_015122272.2 | XP_014977758.1 | 757 | 101 | |||
| XM_015122273.2 | XP_014977759.1 | 785 | 101 | |||
| XM_028838397.1 | XP_028694230.1 | 676 | 101 | |||
| GenBank mRNA | JU321652.1 | - | 723 | 101 | ||
| JU476325.1 | - | 756 | 101 | |||
| JV065388.1 | - | 633 | 101 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RNF126
Genomic Map