Assay Details
| Gene Symbol: DCLRE1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148218.2 | XP_015003704.2 | 3701 | 73 | ||
| XM_015148220.2 | XP_015003706.2 | 3717 | 73 | |||
| GenBank mRNA | JU337393.1 | - | 3018 | 73 | ||
| JV728161.1 | - | 3491 | 73 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DCLRE1A
Genomic Map