Assay Details
| Gene Symbol: STOML2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260882.2 | NP_001247811.1 | 862 | 83 | ||
| GenBank mRNA | JU321476.1 | - | 841 | 83 | ||
| JU472282.1 | - | 858 | 83 | |||
| JV043525.1 | - | 865 | 83 | |||
| JV669581.1 | - | 868 | 83 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

STOML2
Genomic Map