Assay Details
| Gene Symbol: RBM11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001083260.4 | XP_001083260.1 | 174 | 89 | ||
| GenBank mRNA | JV717138.1 | - | 133 | 89 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RBM11
Genomic Map