Assay Details
Gene Symbol: HSDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199126.2 | NP_001186055.1 | 419 | 107 | ||
XM_015116997.2 | XP_014972483.1 | 442 | 107 | |||
XM_028834302.1 | XP_028690135.1 | 442 | 107 | |||
GenBank mRNA | CB310522.1 | - | 236 | 107 | ||
JU336688.1 | - | 256 | 107 | |||
JU473376.1 | - | 266 | 107 | |||
JV045640.1 | - | 270 | 107 | |||
JV724995.1 | - | 268 | 107 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
