Assay Details
Gene Symbol: CASP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260956.1 | NP_001247885.1 | 303 | 126 | ||
XM_028832625.1 | XP_028688458.1 | 342 | 126 | |||
GenBank mRNA | JU473387.1 | - | 303 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
