Assay Details
| Gene Symbol: TCF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114951.1 | NP_001108423.1 | 792 | 82 | ||
| XM_015135905.2 | XP_014991391.2 | 1501 | 82 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TCF19| Gene Symbol: TCF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114951.1 | NP_001108423.1 | 792 | 82 | ||
| XM_015135905.2 | XP_014991391.2 | 1501 | 82 | |||
Set Membership: |
|