Assay Details
| Gene Symbol: KCNK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126335.2 | XP_014981821.1 | 1077 | 99 | ||
| GenBank mRNA | JU327405.1 | - | 329 | 99 | ||
| JV544228.1 | - | 331 | 99 | |||
| JV668806.1 | - | 421 | 99 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

KCNK1
Genomic Map