Assay Details
| Gene Symbol: MRPL53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194467.2 | NP_001181396.1 | 279 | 108 | ||
| GenBank mRNA | JU332831.1 | - | 295 | 108 | ||
| JU472593.1 | - | 322 | 108 | |||
| JV045571.1 | - | 273 | 108 | |||
| JV453055.1 | - | 251 | 108 | |||
| JV490947.1 | - | 279 | 108 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MRPL53
Genomic Map