Assay Details
| Gene Symbol: PSMA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257562.1 | NP_001244491.1 | 197 | 89 | ||
| XM_028850401.1 | XP_028706234.1 | 214 | 89 | |||
| GenBank mRNA | JU335509.1 | - | 197 | 89 | ||
| JU476450.1 | - | 183 | 89 | |||
| JV047690.1 | - | 176 | 89 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PSMA4
Genomic Map