Assay Details
| Gene Symbol: FAM3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132846.2 | XP_014988332.1 | 698 | 78 | ||
| XM_015132847.2 | XP_014988333.1 | 695 | 78 | |||
| XM_015132848.2 | XP_014988334.1 | 998 | 78 | |||
| GenBank mRNA | JU476051.1 | - | 757 | 78 | ||
| JV654006.1 | - | 350 | 78 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FAM3B
Genomic Map