Assay Details
| Gene Symbol: TMEM143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124460.2 | XP_014979946.2 | 1012 | 65 | ||
| XM_015124461.2 | XP_014979947.2 | 907 | 65 | |||
| GenBank mRNA | JV703926.1 | - | 706 | 65 | ||
More Information
Set Membership: |
|

TMEM143
Genomic Map