Assay Details
| Gene Symbol: LSM14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257954.1 | NP_001244883.1 | 1942 | 138 | ||
| XM_015123515.2 | XP_014979001.1 | 2051 | 138 | |||
| GenBank mRNA | JU321516.1 | - | 1942 | 138 | ||
| JU321517.1 | - | 1885 | 138 | |||
| JV044126.1 | - | 2036 | 138 | |||
| JV044127.1 | - | 1979 | 138 | |||
More Information
Set Membership: |
|

LSM14A
Genomic Map