Assay Details
| Gene Symbol: CRNN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001109196.4 | XP_001109196.3 | 59 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001109196.4 transcripts.
Set Membership: |
|

CRNN
Genomic Map