Assay Details
Gene Symbol: GSKIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194191.1 | NP_001181120.1 | 311 | 123 | ||
GenBank mRNA | JU334913.1 | - | 266 | 123 | ||
JU334914.1 | - | 266 | 123 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
