Assay Details
| Gene Symbol: MEX3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121992.2 | XP_014977478.2 | 1320 | 113 | ||
| GenBank mRNA | JU487815.1 | - | 245 | 113 | ||
| JV062838.1 | - | 684 | 113 | |||
More Information
Set Membership: |
|

MEX3C
Genomic Map