Assay Details
| Gene Symbol: FBXO34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266246.1 | NP_001253175.1 | 529 | 156 | ||
| XM_028850219.1 | XP_028706052.1 | 588 | 156 | |||
| XM_028850220.1 | XP_028706053.1 | 565 | 156 | |||
| XM_028850221.1 | XP_028706054.1 | 535 | 156 | |||
| GenBank mRNA | JU335210.1 | - | 569 | 156 | ||
| JU476182.1 | - | 477 | 156 | |||
| JV047489.1 | - | 529 | 156 | |||
| JV549341.1 | - | 597 | 156 | |||
More Information
Set Membership: |
|

FBXO34
Genomic Map