Assay Details
| Gene Symbol: RNF186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001094241.4 | XP_001094241.2 | 143 | 133 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001094241.4 transcripts.
Set Membership: |
|

RNF186
Genomic Map