Assay Details
| Gene Symbol: ZBTB26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116860.2 | XP_014972346.1 | 157 | 77 | ||
| GenBank mRNA | JU320681.1 | - | 68 | 77 | ||
| JU472142.1 | - | 60 | 77 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015116860.2 transcripts.
Set Membership: |
|

ZBTB26
Genomic Map