Assay Details
| Gene Symbol: ZNF711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127750.2 | XP_014983236.1 | 611 | 78 | ||
| XM_015127752.2 | XP_014983238.1 | 669 | 78 | |||
| XM_015127753.2 | XP_014983239.1 | 500 | 78 | |||
| XM_015127754.2 | XP_014983240.1 | 412 | 78 | |||
| XM_015127755.2 | XP_014983241.1 | 611 | 78 | |||
| XM_015127756.2 | XP_014983242.1 | 412 | 78 | |||
| XM_028842071.1 | XP_028697904.1 | 1230 | 78 | |||
| GenBank mRNA | JU319896.1 | - | 394 | 78 | ||
| JU470586.1 | - | 118 | 78 | |||
| JV044915.1 | - | 267 | 78 | |||
More Information
Set Membership: |
|

ZNF711
Genomic Map