Assay Details
| Gene Symbol: IL25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107906.4 | XP_001107906.2 | 410 | 88 | ||
| XM_015143409.2 | XP_014998895.1 | 2617 | 88 | |||
| GenBank mRNA | JU347283.1 | - | 375 | 88 | ||
More Information
Set Membership: |
|

IL25
Genomic Map