Assay Details
| Gene Symbol: ARSJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001096903.4 | XP_001096903.1 | 796 | 68 | ||
| XM_015139124.2 | XP_014994610.1 | 796 | 68 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARSJ| Gene Symbol: ARSJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001096903.4 | XP_001096903.1 | 796 | 68 | ||
| XM_015139124.2 | XP_014994610.1 | 796 | 68 | |||
Set Membership: |
|