Assay Details
| Gene Symbol: MTERF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193943.3 | NP_001180872.3 | 104 | 46 | ||
| XM_015152646.2 | XP_015008132.2 | 199 | 46 | |||
| XM_015152647.2 | XP_015008133.2 | 199 | 46 | |||
| XM_015152648.2 | XP_015008134.2 | 613 | 46 | |||
| XM_028828864.1 | XP_028684697.1 | 203 | 46 | |||
| XM_028828865.1 | XP_028684698.1 | 613 | 46 | |||
| GenBank mRNA | JU323000.1 | - | 153 | 46 | ||
| JU472899.1 | - | 146 | 46 | |||
| JV044504.1 | - | 146 | 46 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001193943.3, XM_015152646.2, XM_015152647.2, XM_028828864.1 transcripts.
Set Membership: |
|

MTERF2
Genomic Map