Assay Details
| Gene Symbol: GPR63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001100856.4 | XP_001100856.2 | 209 | 153 | ||
| GenBank mRNA | JU473681.1 | - | 170 | 153 | ||
| JV704423.1 | - | 206 | 153 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001100856.4 transcripts.
Set Membership: |
|

GPR63
Genomic Map