Assay Details
| Gene Symbol: CHST9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261472.1 | NP_001248401.1 | 292 | 119 | ||
| GenBank mRNA | JU336606.1 | - | 292 | 119 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHST9| Gene Symbol: CHST9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261472.1 | NP_001248401.1 | 292 | 119 | ||
| GenBank mRNA | JU336606.1 | - | 292 | 119 | ||
Set Membership: |
|