Assay Details
| Gene Symbol: TMUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194233.1 | NP_001181162.1 | 471 | 68 | ||
| XM_015135206.2 | XP_014990692.1 | 551 | 68 | |||
| XM_028845037.1 | XP_028700870.1 | 488 | 68 | |||
| GenBank mRNA | JU321029.1 | - | 512 | 68 | ||
| JU473273.1 | - | 484 | 68 | |||
| JV044387.1 | - | 707 | 68 | |||
| JV559649.1 | - | 461 | 68 | |||
More Information
Set Membership: |
|

TMUB1
Genomic Map