Assay Details
| Gene Symbol: MAFG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194569.3 | NP_001181498.1 | 250 | 69 | ||
| XM_028835692.1 | XP_028691525.1 | 252 | 69 | |||
| GenBank mRNA | JU320800.1 | - | 249 | 69 | ||
| JU472492.1 | - | 117 | 69 | |||
| JV043587.1 | - | 117 | 69 | |||
More Information
Set Membership: |
|

MAFG
Genomic Map