Assay Details
| Gene Symbol: FAM222A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194323.2 | NP_001181252.1 | 272 | 133 | ||
| XM_028828908.1 | XP_028684741.1 | 201 | 133 | |||
| GenBank mRNA | JU322508.1 | - | 643 | 133 | ||
| JV602144.1 | - | 377 | 133 | |||
| JV637762.1 | - | 526 | 133 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194323.2, XM_028828908.1 transcripts.
Set Membership: |
|

FAM222A
Genomic Map