Assay Details
| Gene Symbol: MRPS33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266016.1 | NP_001252945.1 | 373 | 151 | ||
| XM_015135058.2 | XP_014990544.1 | 381 | 151 | |||
| XM_028844777.1 | XP_028700610.1 | 271 | 151 | |||
| GenBank mRNA | JU323312.1 | - | 360 | 151 | ||
| JU473610.1 | - | 264 | 151 | |||
| JV045176.1 | - | 373 | 151 | |||
| JV601070.1 | - | 239 | 151 | |||
More Information
Set Membership: |
|

MRPS33
Genomic Map