Assay Details
| Gene Symbol: ASB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001097027.4 | XP_001097027.2 | 1010 | 117 | ||
| XM_015127577.2 | XP_014983063.1 | 931 | 117 | |||
More Information
Set Membership: |
|

ASB12
Genomic Map