Assay Details
| Gene Symbol: ATOH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015147518.2 | XP_015003004.1 | 266 | 139 | ||
| GenBank mRNA | JV709491.1 | - | 182 | 139 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015147518.2 transcripts.
Set Membership: |
|

ATOH7
Genomic Map