Assay Details
| Gene Symbol: CCDC89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115435.2 | XP_014970921.2 | 62 | 123 | ||
| XM_028832892.1 | XP_028688725.1 | 62 | 123 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015115435.2, XM_028832892.1 transcripts.
Set Membership: |
|

CCDC89
Genomic Map