Assay Details
| Gene Symbol: PRR18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028846984.1 | XP_028702817.1 | 825 | 139 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028846984.1 transcripts.
Set Membership: |
|

PRR18
Genomic Map