Assay Details
Gene Symbol: ERAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194200.2 | NP_001181129.1 | 1213 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
