Assay Details
Gene Symbol: TICAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130428.1 | NP_001123900.1 | 391 | 80 | ||
XM_015122534.2 | XP_014978020.2 | 80 | ||||
GenBank mRNA | AB446490.1 | - | 391 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
