Assay Details
| Gene Symbol: CPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265884.1 | NP_001252813.1 | 263 | 85 | ||
| GenBank mRNA | JU332983.1 | - | 263 | 85 | ||
| JV636857.1 | - | 300 | 85 | |||
More Information
Set Membership: |
|

CPT2
Genomic Map