Assay Details
| Gene Symbol: SGCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001113209.4 | XP_001113209.4 | 1219 | 138 | ||
| XM_015141314.2 | XP_014996800.2 | 1173 | 138 | |||
| XM_028849861.1 | XP_028705694.1 | 832 | 138 | |||
More Information
Set Membership: |
|

SGCD
Genomic Map