Assay Details
| Gene Symbol: SERPINA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001088790.4 | XP_001088790.3 | 729 | 132 | ||
| XM_015127950.2 | XP_014983436.1 | 729 | 132 | |||
| XM_015127951.2 | XP_014983437.1 | 729 | 132 | |||
More Information
Set Membership: |
|

SERPINA7
Genomic Map