Assay Details
| Gene Symbol: CYP27A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194021.1 | NP_001180950.1 | 1917 | 70 | ||
| GenBank mRNA | JU322833.1 | - | 1921 | 70 | ||
| JV043738.1 | - | 1910 | 70 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CYP27A1
Genomic Map