Assay Details
| Gene Symbol: FCRLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001118137.3 | XP_001118137.1 | 287 | 134 | ||
| XM_015113982.1 | XP_014969468.1 | 67 | 134 | |||
| XM_015113997.1 | XP_014969483.1 | 67 | 134 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FCRLB
Genomic Map