Assay Details
| Gene Symbol: HCST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032835.2 | NP_001028007.1 | 64 | 78 | ||
| XM_015123559.2 | XP_014979045.2 | 147 | 78 | |||
| XM_015123560.2 | XP_014979046.2 | 147 | 78 | |||
| XM_028838319.1 | XP_028694152.1 | 846 | 78 | |||
| GenBank mRNA | AF321610.1 | - | 64 | 78 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HCST
Genomic Map