Assay Details
| Gene Symbol: GTPBP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194320.3 | NP_001181249.3 | 659 | 93 | ||
| XM_001105772.4 | XP_001105772.3 | 599 | 93 | |||
| XM_015129810.2 | XP_014985296.2 | 731 | 93 | |||
| GenBank mRNA | JV713074.1 | - | 653 | 93 | ||
More Information
Set Membership: |
|

GTPBP8
Genomic Map