Assay Details
| Gene Symbol: AKAP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193725.1 | NP_001180654.1 | 178 | 152 | ||
| XM_015128098.2 | XP_014983584.1 | 437 | 152 | |||
| XM_015128100.2 | XP_014983586.1 | 415 | 152 | |||
More Information
Set Membership: |
|

AKAP14
Genomic Map